Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nqo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163239.1 | NP_001156711.1 | 4 - 5 | 547 | 82 | |
NM_001163241.1 | NP_001156713.1 | 5 - 6 | 635 | 82 | ||
NM_001163242.1 | NP_001156714.1 | 5 - 6 | 564 | 82 | ||
NM_020282.3 | NP_064678.1 | 6 - 7 | 717 | 82 | ||
GenBank mRNA | AF252260.1 | - | 5 - 6 | 563 | 82 | |
AK008138.1 | - | 4 - 5 | 519 | 82 | ||
AK008465.1 | - | 4 - 5 | 522 | 82 | ||
AK010842.1 | - | 5 - 6 | 607 | 82 | ||
AK049781.1 | - | 5 - 6 | 573 | 82 | ||
BC027629.1 | - | 6 - 7 | 625 | 1225122 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |