Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2187829 |
AK014187.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
198855 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
276403 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
23082 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
933340 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
642712 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
589218 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
800577 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
459238 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
85562 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
844645 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
497457 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
241202 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
614659 |
AK005665.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
614659 |
AY600990.2 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
614659 |
BC048681.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
614659 |
NM_027024.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
857471 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
323517 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
788638 |
AK039417.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
132083 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
129836 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
76826 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
86094 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P158964 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P158964 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P158964 |
- |
- |