Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024465.3 | NP_077785.2 | 9 - 10 | 1051 | 103 | |
GenBank mRNA | AK178612.1 | - | 2 - 3 | 121 | 103 | |
AK185693.1 | - | 2 - 3 | 99 | 103 | ||
AK198485.1 | - | 2 - 3 | 121 | 103 | ||
AK202769.1 | - | 2 - 3 | 121 | 103 | ||
AK217770.1 | - | 2 - 3 | 121 | 103 | ||
BC002138.1 | - | 9 - 10 | 1020 | 103 | ||
BC002263.1 | - | 5 - 6 | 354 | 3495118 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |