Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suv39h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022724.4 | NP_073561.2 | 5 - 6 | 1371 | 74 | |
NR_027509.1 | 4 - 5 | 1225 | 74 | |||
GenBank mRNA | AF149205.1 | - | 5 - 6 | 1349 | 74 | |
AK015728.1 | - | 3 - 4 | 417 | 74 | ||
AK045206.1 | - | 4 - 5 | 1209 | 74 | ||
AK083457.1 | - | 5 - 6 | 1349 | 74 | ||
AK165278.1 | - | 4 - 5 | 1024 | 74 | ||
BC032960.1 | - | 4 - 5 | 694 | 1330053 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |