Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20962886 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20962881 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20962880 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20962882 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20962884 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20962883 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20962885 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P361551 |
U08215.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P361551 |
BC002056.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P361551 |
AK031489.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P361551 |
AK145937.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P361551 |
NM_015765.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P361551 |
U08215.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P361551 |
BC002056.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P361551 |
AK031489.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P361551 |
AK145937.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P361551 |
NM_015765.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
933702 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
16066 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
397985 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
244656 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
742594 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
50228 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
147053 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316667 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
409554 |
AK019818.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
409554 |
XM_001003948.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
409554 |
XM_001472902.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
429419 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
427124 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
918029 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
533448 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
79726 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
83088 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
148463 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P148606 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P134988 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P166092 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P361551 |
U08215.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P361551 |
BC002056.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P361551 |
AK031489.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P361551 |
AK145937.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P361551 |
NM_015765.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P361551 |
U08215.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P361551 |
BC002056.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P361551 |
AK031489.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P361551 |
AK145937.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P361551 |
NM_015765.2 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04411164 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P134988 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04411211 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P166092 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P148606 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P361551 |
U08215.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P361551 |
BC002056.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P361551 |
AK031489.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P361551 |
AK145937.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P361551 |
NM_015765.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P361551 |
U08215.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P361551 |
BC002056.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P361551 |
AK031489.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P361551 |
AK145937.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P361551 |
NM_015765.2 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P113730 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P124317 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P166092 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P134988 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P148606 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01352718 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01352757 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01352712 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01352733 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20962882 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20962885 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20962886 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20962883 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20962880 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20962884 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20962881 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962801 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962734 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962833 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962776 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962827 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962761 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962719 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962824 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962768 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962870 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962773 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962834 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962832 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962856 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962809 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962716 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962752 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962843 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962797 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962858 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962865 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962871 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962875 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962825 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962699 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962868 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962829 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962764 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962755 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962811 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962777 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962830 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962792 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962796 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962823 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962775 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962850 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962766 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962806 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962814 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962780 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962869 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962711 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962714 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962784 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962862 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962880 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962883 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962807 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962853 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962877 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962750 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962884 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962816 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962712 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962692 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962791 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962689 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962795 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962886 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962860 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962770 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962820 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962866 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962874 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962759 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962788 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20962757 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962830 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01352738 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962851 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01352757 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962734 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01867254 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962848 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962862 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962869 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962858 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962824 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962853 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962873 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962740 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962742 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962846 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962841 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962866 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962874 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962825 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962884 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02243092 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962843 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962829 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962870 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962865 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962746 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01695116 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962737 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962840 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01372369 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962860 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962744 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962877 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01695110 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962875 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962827 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962856 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01352739 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01846299 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962736 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962834 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20962883 |
- |
- |