Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015765.2 | NP_056580.2 | 5 - 6 | 507 | 85 | |
GenBank mRNA | AK031489.1 | - | 446 | 85 | ||
AK145937.1 | - | 5 - 6 | 461 | 85 | ||
AK153397.1 | - | 5 - 6 | 469 | 85 | ||
BC002056.1 | - | 5 - 6 | 435 | 3493119 | 85 | |
BU505607.1 | - | 2 - 3 | 158 | 85 | ||
U08215.1 | - | 5 - 6 | 467 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |