Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200023745.mm.1 |
NM_026045.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023745.mm.1 |
NM_026045.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023745.mm.1 |
TC0200002863.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023748.mm.1 |
NM_026045.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023748.mm.1 |
NM_026045.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023748.mm.1 |
TC0200002863.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023743.mm.1 |
NM_026045.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023743.mm.1 |
NM_026045.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023743.mm.1 |
TC0200002863.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023744.mm.1 |
NM_026045.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023744.mm.1 |
NM_026045.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023744.mm.1 |
TC0200002863.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023741.mm.1 |
NM_026045.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023741.mm.1 |
NM_026045.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023741.mm.1 |
TC0200002863.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023742.mm.1 |
NM_026045.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023742.mm.1 |
NM_026045.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023742.mm.1 |
TC0200002863.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023750.mm.1 |
NM_026045.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023750.mm.1 |
NM_026045.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023750.mm.1 |
TC0200002863.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023749.mm.1 |
NM_026045.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023749.mm.1 |
NM_026045.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023749.mm.1 |
TC0200002863.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023746.mm.1 |
NM_026045.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023746.mm.1 |
NM_026045.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023746.mm.1 |
TC0200002863.mm.1 |
MTA TC |