Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028716.4 | NP_082992.1 | 6 - 7 | 651 | 71 | |
GenBank mRNA | AK014030.1 | - | 5 - 6 | 1071 | 71 | |
AK014380.1 | - | 6 - 7 | 652 | 71 | ||
BC027776.1 | - | 6 - 7 | 646 | 5352251 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |