Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080554.2 | NP_542121.2 | 6 - 7 | 852 | 71 | |
GenBank mRNA | AK005335.1 | - | 6 - 7 | 830 | 71 | |
AK011026.1 | - | 6 - 7 | 829 | 72 | ||
AK029067.1 | - | 5 - 6 | 1149 | 71 | ||
AK032992.1 | - | 6 - 7 | 830 | 71 | ||
AK078419.1 | - | 6 - 7 | 830 | 71 | ||
AK145587.1 | - | 6 - 7 | 829 | 71 | ||
AK148284.1 | - | 853 | 71 | |||
AK205476.1 | - | 1 - 2 | 153 | 71 | ||
AK207702.1 | - | 1 - 2 | 153 | 71 | ||
AK220321.1 | - | 6 - 7 | 793 | 71 | ||
BC019112.1 | - | 6 - 7 | 863 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |