Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290564.1 | NP_001277493.1 | 2 - 3 | 404 | 61 | |
GenBank mRNA | AK028040.1 | - | 2 - 3 | 403 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |