Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081310.2 | NP_001074779.1 | 1 - 2 | 318 | 96 | |
GenBank mRNA | AK144489.1 | - | 1 - 2 | 319 | 96 | |
BY733764.1 | - | 1 - 2 | 319 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |