Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177268.4 | NP_796242.2 | 2 - 3 | 1373 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |