Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dolpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290508.1 | NP_001277437.1 | 1 - 2 | 262 | 59 | |
GenBank mRNA | AK002544.1 | - | 1 - 2 | 101 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |