Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290484.1 | NP_001277413.1 | 1 - 2 | 452 | 75 | |
NM_148928.3 | NP_683730.1 | 1 - 2 | 452 | 75 | ||
GenBank mRNA | AK137359.1 | - | 1 - 2 | 441 | 75 | |
AK147014.1 | - | 1 - 2 | 430 | 75 | ||
BC027055.1 | - | 1 - 2 | 419 | 5360397 | 75 | |
BC027247.1 | - | 1 - 2 | 362 | 2631223 | 75 | |
BC039171.1 | - | 1 - 2 | 373 | 4500799 | 75 | |
BQ179286.1 | - | 1 - 2 | 452 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |