Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145528.3 | NP_663503.1 | 9 - 10 | 1056 | 90 | |
GenBank mRNA | AK122339.1 | - | 5 - 6 | 775 | 90 | |
AK149672.1 | - | 9 - 10 | 1015 | 90 | ||
AK154470.1 | - | 9 - 10 | 1044 | 90 | ||
AK163962.1 | - | 9 - 10 | 1010 | 90 | ||
AK171206.1 | - | 9 - 10 | 1042 | 90 | ||
AK171440.1 | - | 9 - 10 | 1033 | 90 | ||
BC016669.1 | - | 9 - 10 | 1040 | 2648797 | 90 | |
BC023673.1 | - | 9 - 10 | 1006 | 5343236 | 90 | |
BC023712.1 | - | 9 - 10 | 1029 | 5346303 | 90 | |
BC033419.1 | - | 9 - 10 | 1007 | 5359944 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |