Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145813.1 | NP_001139285.1 | 5 - 6 | 502 | 67 | |
NM_010125.3 | NP_034255.2 | 5 - 6 | 593 | 67 | ||
GenBank mRNA | AF049702.1 | - | 5 - 6 | 584 | 67 | |
AK132484.1 | - | 5 - 6 | 913 | 67 | ||
AK145214.1 | - | 5 - 6 | 600 | 67 | ||
BC012424.1 | - | 5 - 6 | 535 | 3584025 | 67 | |
BC021629.1 | - | 5 - 6 | 546 | 2651428 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |