Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009804.2 | NP_033934.2 | 2 - 3 | 351 | 66 | |
GenBank mRNA | AK075853.1 | - | 2 - 3 | 321 | 66 | |
AK088168.1 | - | 2 - 3 | 313 | 66 | ||
AK128919.1 | - | 2 - 3 | 326 | 66 | ||
AK133881.1 | - | 2 - 3 | 319 | 66 | ||
AK148961.1 | - | 2 - 3 | 319 | 66 | ||
AK150893.1 | - | 2 - 3 | 322 | 66 | ||
AK159152.1 | - | 2 - 3 | 322 | 66 | ||
AK159885.1 | - | 2 - 3 | 322 | 66 | ||
AK159891.1 | - | 2 - 3 | 273 | 66 | ||
AK159913.1 | - | 2 - 3 | 303 | 66 | ||
AK169069.1 | - | 2 - 3 | 316 | 66 | ||
AK188260.1 | - | 1 - 2 | 35 | 66 | ||
AK188401.1 | - | 1 - 2 | 35 | 66 | ||
AY040626.1 | - | 2 - 3 | 292 | 66 | ||
BC013447.1 | - | 2 - 3 | 308 | 4241333 | 66 | |
BC047126.1 | - | 2 - 3 | 266 | 66 | ||
L25069.1 | - | 2 - 3 | 323 | 66 | ||
M62897.1 | - | 2 - 3 | 236 | 66 | ||
X52108.1 | - | 2 - 3 | 236 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |