Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cage1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027724.2 | NP_082000.2 | 7 - 8 | 2290 | 123 | |
GenBank mRNA | AK132733.1 | - | 3 - 4 | 838 | 123 | |
AY695827.1 | - | 6 - 7 | 2025 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |