Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159640.2 | NP_001153112.1 | 8 - 9 | 791 | 61 | |
NM_001159641.2 | NP_001153113.1 | 8 - 9 | 791 | 61 | ||
NM_175225.5 | NP_780434.1 | 8 - 9 | 1058 | 61 | ||
NR_110350.1 | 7 - 8 | 978 | 61 | |||
NR_110351.1 | 8 - 9 | 791 | 61 | |||
NR_110352.1 | 7 - 8 | 722 | 61 | |||
GenBank mRNA | AK015624.1 | - | 8 - 9 | 791 | 61 | |
AK036512.1 | - | 8 - 9 | 1014 | 61 | ||
AK132709.1 | - | 7 - 8 | 708 | 61 | ||
AK146320.1 | - | 8 - 9 | 774 | 61 | ||
AK160983.1 | - | 7 - 8 | 939 | 61 | ||
BC024597.1 | - | 8 - 9 | 778 | 4036044 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |