Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpped2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143683.1 | NP_001137155.1 | 1 - 2 | 363 | 84 | |
GenBank mRNA | AK161666.1 | - | 1 - 2 | 364 | 84 | |
AK165186.1 | - | 1 - 2 | 87 | |||
BC020182.1 | - | 1 - 2 | 273 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |