Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpped2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143683.1 | NP_001137155.1 | 2 - 3 | 612 | 60 | |
NM_029837.1 | NP_084113.1 | 2 - 3 | 325 | 60 | ||
GenBank mRNA | AK012553.1 | - | 2 - 3 | 326 | 60 | |
AK161666.1 | - | 2 - 3 | 613 | 60 | ||
AK165186.1 | - | 2 - 3 | 809 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |