Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110223.1 | NP_001103693.1 | 1 - 2 | 148 | 88 | |
NM_001110224.1 | NP_001103694.1 | 1 - 2 | 148 | 88 | ||
NM_010025.2 | NP_034155.2 | 1 - 2 | 148 | 88 | ||
GenBank mRNA | AB011678.1 | - | 1 - 2 | 244 | 88 | |
AK014264.1 | - | 1 - 2 | 149 | 88 | ||
AK164119.1 | - | 1 - 2 | 35 | 88 | ||
BC056391.1 | - | 1 - 2 | 41 | 6855006 | 88 | |
BC057010.1 | - | 1 - 2 | 133 | 6825750 | 88 | |
BC062974.1 | - | 1 - 2 | 133 | 6825750 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |