Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110222.1 | NP_001103692.1 | 5 - 6 | 1196 | 72 | |
NM_001110223.1 | NP_001103693.1 | 5 - 6 | 1118 | 72 | ||
NM_001110224.1 | NP_001103694.1 | 5 - 6 | 1118 | 72 | ||
GenBank mRNA | AB011678.1 | - | 5 - 6 | 1214 | 72 | |
AF045547.1 | - | 5 - 6 | 999 | 72 | ||
AK048410.1 | - | 5 - 6 | 1124 | 72 | ||
AK164119.1 | - | 5 - 6 | 1005 | 72 | ||
AY560329.1 | - | 5 - 6 | 1006 | 72 | ||
BC056391.1 | - | 5 - 6 | 1011 | 6855006 | 72 | |
BC057010.1 | - | 5 - 6 | 1103 | 6825750 | 72 | |
BC062974.1 | - | 5 - 6 | 1103 | 6825750 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |