Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK007308.1 | - | 4 - 5 | 870 | 101 | |
AK015125.1 | - | 4 - 5 | 846 | 101 | ||
BC043075.1 | - | 5 - 6 | 901 | 6404784 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |