Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289723.1 | NP_001276652.1 | 1 - 2 | 131 | 79 | |
GenBank mRNA | AK031046.1 | - | 1 - 2 | 128 | 79 | |
BY200699.1 | - | 1 - 2 | 132 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |