Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175207.4 | NP_780416.2 | 1 - 2 | 182 | 95 | |
GenBank mRNA | AK032641.1 | - | 1 - 2 | 183 | 95 | |
AK051108.1 | - | 1 - 2 | 202 | 95 | ||
AK172167.1 | - | 1 - 2 | 157 | 95 | ||
BC023687.1 | - | 1 - 2 | 156 | 5343549 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |