Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027660.1 | NP_081936.1 | 6 - 7 | 1254 | 117 | |
GenBank mRNA | AK016718.1 | - | 6 - 7 | 1253 | 117 | |
AY252101.1 | - | 6 - 7 | 1254 | 117 | ||
BC127605.1 | - | 6 - 7 | 1152 | 40057157 | 117 | |
BC128270.1 | - | 6 - 7 | 1152 | 40057161 | 117 | |
BC139074.1 | - | 6 - 7 | 1221 | 8862096 | 117 | |
BC139075.1 | - | 6 - 7 | 1221 | 8862097 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |