Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173030.2 | NP_766618.2 | 1 - 2 | 341 | 123 | |
GenBank mRNA | AK038387.1 | - | 1 - 2 | 342 | 123 | |
AK045417.1 | - | 1 - 2 | 342 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |