Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Accsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033452.4 | NP_001028624.2 | 9 - 10 | 1225 | 103 | |
GenBank mRNA | AK135828.1 | - | 9 - 10 | 1172 | 103 | |
AK142866.1 | - | 7 - 8 | 716 | 103 | ||
AK163963.1 | - | 9 - 10 | 1307 | 103 | ||
BC141227.1 | - | 7 - 8 | 604 | 9055839 | 103 | |
BC145551.1 | - | 7 - 8 | 604 | 103 | ||
BC145552.1 | - | 7 - 8 | 604 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |