Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026944.1 | NP_081220.1 | 4 - 5 | 337 | 63 | |
GenBank mRNA | AK145586.1 | - | 4 - 5 | 723 | 63 | |
BC018196.1 | - | 4 - 5 | 337 | 3993214 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |