Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172262.3 | NP_758466.1 | 18 - 19 | 2305 | 68 | |
GenBank mRNA | AK028553.1 | - | 8 - 9 | 971 | 68 | |
AK078920.1 | - | 7 - 8 | 863 | 68 | ||
BC023917.1 | - | 18 - 19 | 2246 | 5323227 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |