Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Disp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170593.3 | NP_733481.1 | 3 - 4 | 581 | 64 | |
GenBank mRNA | AK036480.1 | - | 562 | 64 | ||
AK038938.1 | - | 3 - 4 | 544 | 64 | ||
AK045560.1 | - | 582 | 64 | |||
AY150699.1 | - | 3 - 4 | 382 | 64 | ||
BC118526.1 | - | 3 - 4 | 376 | 40099214 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |