Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173428.3 | NP_775604.3 | 18 - 19 | 2902 | 77 | |
GenBank mRNA | AJ491857.1 | - | 16 - 17 | 2533 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |