Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081057.2 | NP_001074526.1 | 3 - 4 | 71 | ||
NM_001289510.1 | NP_001276439.1 | 2 - 3 | 71 | |||
NM_175336.3 | NP_780545.2 | 3 - 4 | 71 | |||
GenBank mRNA | AK031736.1 | - | 2 - 3 | 71 | ||
AK129113.1 | - | 2 - 3 | 71 | |||
AK147592.1 | - | 2 - 3 | 71 | |||
BC138354.1 | - | 2 - 3 | 8861374 | 71 | ||
BC145250.1 | - | 2 - 3 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |