Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4921507L20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045827.1 | 1 - 2 | 605 | 80 | ||
GenBank mRNA | AK014837.1 | - | 1 - 2 | 606 | 80 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 13889 | Gm13889 |
Ag2, OTTMUSG00000014994 |
-22.9kb | Anti-sense | Chr. 2 - 93955810 - 93957100 on NCBI Build GRCm38 |
Downstream | hydroxysteroid (17-beta) dehydrogenase 12 | Hsd17b12 |
2610510O05Rik, AI172963, KIK-I, Kik1 |
51.5kb | Anti-sense | Chr. 2 - 94032697 - 94157909 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |