Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271879.1 | NP_001258808.1 | 16 - 17 | 1922 | 95 | |
NM_177694.6 | NP_808362.2 | 17 - 18 | 2072 | 95 | ||
NR_073508.1 | 17 - 18 | 2065 | 95 | |||
GenBank mRNA | AB125740.1 | - | 17 - 18 | 2072 | 95 | |
AB206762.1 | - | 17 - 18 | 2072 | 95 | ||
AK034197.1 | - | 17 - 18 | 2054 | 95 | ||
BC109163.2 | - | 16 - 17 | 1844 | 40047987 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |