Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008569.2 | NP_032595.2 | 10 - 11 | 1520 | 96 | |
GenBank mRNA | AK052404.1 | - | 10 - 11 | 1501 | 96 | |
AK054305.1 | - | 10 - 11 | 1502 | 96 | ||
AK077847.1 | - | 10 - 11 | 1521 | 96 | ||
AK090134.1 | - | 10 - 11 | 1503 | 96 | ||
AK149762.1 | - | 10 - 11 | 1492 | 96 | ||
AK164804.1 | - | 10 - 11 | 1506 | 96 | ||
BC139002.1 | - | 10 - 11 | 1501 | 8862024 | 96 | |
BC145492.1 | - | 10 - 11 | 1501 | 9054055 | 96 | |
X80169.1 | - | 10 - 11 | 1505 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |