Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem87b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028248.2 | NP_082524.2 | 7 - 8 | 684 | 94 | |
GenBank mRNA | AK011959.1 | - | 7 - 8 | 605 | 94 | |
AK050203.1 | - | 7 - 8 | 676 | 94 | ||
AK171811.1 | - | 8 - 9 | 600 | 94 | ||
AK172611.1 | - | 8 - 9 | 600 | 94 | ||
BC031488.1 | - | 7 - 8 | 640 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |