Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011812.4 | NP_035942.1 | 3 - 4 | 487 | 75 | |
GenBank mRNA | AF112151.1 | - | 3 - 4 | 437 | 75 | |
AK084895.1 | - | 3 - 4 | 328 | 75 | ||
AK085170.1 | - | 3 - 4 | 317 | 75 | ||
AK090129.1 | - | 3 - 4 | 426 | 75 | ||
AK159189.1 | - | 3 - 4 | 250 | 75 | ||
AK159471.1 | - | 3 - 4 | 470 | 75 | ||
AK159960.1 | - | 3 - 4 | 457 | 75 | ||
AK165018.1 | - | 3 - 4 | 280 | 75 | ||
BC006636.1 | - | 3 - 4 | 177 | 3482574 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |