Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhtkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081131.2 | NP_001074600.1 | 9 - 10 | 1882 | 54 | |
GenBank mRNA | AK049272.1 | - | 1873 | 54 | ||
AK050057.1 | - | 191 | 54 | |||
BC117994.1 | - | 1833 | 40097991 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |