Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136081.2 | NP_001129553.1 | 15 - 16 | 2070 | 108 | |
NM_182990.4 | NP_892035.2 | 15 - 16 | 2199 | 108 | ||
GenBank mRNA | AK146446.1 | - | 15 - 16 | 2122 | 108 | |
AK146585.1 | - | 15 - 16 | 2119 | 108 | ||
AK146607.1 | - | 15 - 16 | 2102 | 108 | ||
AK146691.1 | - | 15 - 16 | 2101 | 108 | ||
AK147005.1 | - | 15 - 16 | 2130 | 108 | ||
AK158552.1 | - | 15 - 16 | 2070 | 108 | ||
BC096682.1 | - | 1 - 2 | 106 | 108 | ||
S50213.1 | - | 14 - 15 | 1944 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |