Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200025856.mm.1 |
NM_001082476.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025856.mm.1 |
NM_001082476.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025856.mm.1 |
NM_001252541.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025856.mm.1 |
NM_001252541.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025856.mm.1 |
NM_001252542.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025856.mm.1 |
NM_001252542.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025856.mm.1 |
TC0200003140.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200025857.mm.1 |
NM_001082476.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025857.mm.1 |
NM_001082476.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025857.mm.1 |
NM_001252541.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025857.mm.1 |
NM_001252541.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025857.mm.1 |
NM_001252542.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025857.mm.1 |
NM_001252542.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025857.mm.1 |
TC0200003140.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200025852.mm.1 |
NM_001082476.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025852.mm.1 |
NM_001082476.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025852.mm.1 |
NM_001252541.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025852.mm.1 |
NM_001252541.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025852.mm.1 |
NM_001252542.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025852.mm.1 |
NM_001252542.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025852.mm.1 |
TC0200003140.mm.1 |
MTA TC |