Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009849.2 | NP_033979.2 | 7 - 8 | 1248 | 61 | |
GenBank mRNA | AF042811.1 | - | 7 - 8 | 1172 | 61 | |
AK002553.1 | - | 7 - 8 | 1220 | 61 | ||
AY376711.1 | - | 7 - 8 | 1181 | 61 | ||
BC011241.1 | - | 7 - 8 | 1180 | 3582550 | 61 | |
U91511.1 | - | 5 - 6 | 1135 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |