Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009157.4 | NP_033183.1 | 9 - 10 | 1085 | 95 | |
GenBank mRNA | AK036928.1 | - | 9 - 10 | 1089 | 95 | |
AK044534.1 | - | 9 - 10 | 1077 | 95 | ||
AK044698.1 | - | 9 - 10 | 1077 | 95 | ||
AK139777.1 | - | 10 - 11 | 1108 | 95 | ||
BC029833.1 | - | 7 - 8 | 836 | 3962163 | 95 | |
U18310.1 | - | 9 - 10 | 1060 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |