Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kynu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289593.1 | NP_001276522.1 | 5 - 6 | 534 | 94 | |
NM_001289594.1 | NP_001276523.1 | 5 - 6 | 534 | 94 | ||
NM_027552.2 | NP_081828.1 | 5 - 6 | 534 | 94 | ||
GenBank mRNA | AK014484.1 | - | 5 - 6 | 530 | 94 | |
AK040584.1 | - | 5 - 6 | 509 | 94 | ||
AK050024.1 | - | 5 - 6 | 531 | 94 | ||
BC026950.1 | - | 5 - 6 | 505 | 5136970 | 94 | |
BC069848.1 | - | 5 - 6 | 540 | 5097145 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |