Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025664.5 | NP_079940.2 | 12 - 13 | 1441 | 71 | |
GenBank mRNA | AK139802.1 | - | 12 - 13 | 1475 | 71 | |
AK149739.1 | - | 12 - 13 | 1455 | 71 | ||
AK149849.1 | - | 12 - 13 | 1495 | 71 | ||
AK155830.1 | - | 1476 | 71 | |||
AK157517.1 | - | 12 - 13 | 1457 | 71 | ||
AK170339.1 | - | 1457 | 71 | |||
AK204095.1 | - | 3 - 4 | 71 | |||
BC014814.1 | - | 12 - 13 | 1441 | 3257422 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |