Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025664.5 | NP_079940.2 | 4 - 5 | 449 | 92 | |
GenBank mRNA | AK139802.1 | - | 4 - 5 | 483 | 92 | |
AK149739.1 | - | 4 - 5 | 463 | 92 | ||
AK149849.1 | - | 4 - 5 | 503 | 92 | ||
AK155830.1 | - | 484 | 92 | |||
AK170339.1 | - | 465 | 92 | |||
AK194036.1 | - | 1 - 2 | 113 | 92 | ||
AK202482.1 | - | 1 - 2 | 127 | 92 | ||
BC014814.1 | - | 4 - 5 | 449 | 3257422 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |