Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289488.1 | NP_001276417.1 | 1 - 2 | 81 | 63 | |
GenBank mRNA | AK004674.1 | - | 1 - 2 | 81 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |