Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029309.2 | NP_083585.1 | 1 - 2 | 97 | 79 | |
GenBank mRNA | AK005808.1 | - | 1 - 2 | 85 | 79 | |
BC036965.1 | - | 1 - 2 | 103 | 4511210 | 79 | |
BC048673.1 | - | 1 - 2 | 97 | 6774106 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |