Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029774.2 | NP_084050.1 | 2 - 3 | 550 | 78 | |
GenBank mRNA | AK016577.1 | - | 2 - 3 | 223 | 78 | |
AM690755.1 | - | 2 - 3 | 550 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |